accessibility menu, dialog, popup

UserName

New Feature

You can now browse the site in English or Français.

CAS RN 78-27-3

HOCH

CAS RN 78-27-3

IUPAC Name: 1-ethynylcyclohexan-1-ol
Synonyms: 1-ethynyl cyclohexanol
Molecular Weight (g/mol): 124.18
Molecular Formula: C8H12O
InChi Key: QYLFHLNFIHBCPR-UHFFFAOYSA-N
SMILES: OC1(CCCCC1)C#C
Molecular Weight (g/mol) 
  • (5)
  • (2)
Quantity 
  • (1)
  • (3)
  • (1)
  • (2)
Boiling Point 
  • (2)
  • (3)
Percent Purity 
  • (2)
  • (3)
Physical Form 
  • (3)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (1)
  • (1)
  • (2)
  • (2)
  • (2)
  • (5)
  • (2)
  • (1)
  • (3)
  • (1)
  • (2)
  • (2)
  • (3)
  • (2)
  • (3)
  • (3)