accessibility menu, dialog, popup

UserName

New Feature

You can now browse the site in English or Français.

CAS RN 78415-72-2

CH3NOHNN

CAS RN 78415-72-2

IUPAC Name: 2-methyl-6-oxo-1,6-dihydro-[3,4'-bipyridine]-5-carbonitrile
Synonyms: milrinone corotrope milrila corotrop
Molecular Weight (g/mol): 211.22
Molecular Formula: C12H9N3O
InChi Key: PZRHRDRVRGEVNW-UHFFFAOYSA-N
SMILES: CC1=C(C=C(C#N)C(=O)N1)C1=CC=NC=C1
Molecular Weight (g/mol) 
  • (4)
  • (4)
Percent Purity 
  • (2)
  • (2)
  • (2)
Quantity 
  • (1)
  • (1)
  • (1)
  • (2)
  • (1)
Certifications/Compliance 
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (1)
  • (1)
  • (2)
  • (2)
  • (2)
  • (2)
  • (4)
  • (4)
  • (2)
  • (2)
  • (2)
  • (1)
  • (1)
  • (1)
  • (2)
  • (1)
  • (1)
  • (1)
  • (2)
  • (1)