missing translation for 'onlineSavingsMsg'
Learn More

Milrinone, 98+%, Thermo Scientific™

Potent cAMP-specific phosphodiesterase inhibitor

Brand:  Thermo Scientific Alfa Aesar J62659.MF

Quantity 50mg

 View more versions of this product

Product Code. 15444989

  • ONLINE EXCLUSIVE PRICE
    506.00€
    496.00€ / 50mg
    Save 10.00€
    1% Off

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Description

Description

This Thermo Scientific brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.

Applications
It is used (particularly intravenously, as the lactate) for the short-term management of severe heart failure. Milrinone inhibits the action of phosphodiesterase-3 preventing degradation of cAMP. The corresponding increase in cAMP levels leads to increased activation of protein kinase A. It is a selective phosphodiesterase inhibitor with vasodilating and positive inotropic activity.

Solubility
Soluble in DMSO. Insoluble in water.

Notes
Store away from oxidizing agents. Keep the container tightly closed and place it in a cool, dry and well ventilated condition.
TRUSTED_SUSTAINABILITY
Specifications

Chemical Identifiers

78415-72-2
211.224
PZRHRDRVRGEVNW-UHFFFAOYSA-N
4197
6-methyl-2-oxo-5-pyridin-4-yl-1H-pyridine-3-carbonitrile
C12H9N3O
MFCD00133539
milrinone, primacor, corotrope, corotrop, milrinona, milrinonum, milrinonum latin, milrinona spanish, milrila, unii-ju9yax04c7
CHEBI:50693
CC1=C(C=C(C(=O)N1)C#N)C2=CC=NC=C2

Specifications

78415-72-2
MFCD00133539
UN2811
milrinone, primacor, corotrope, corotrop, milrinona, milrinonum, milrinonum latin, milrinona spanish, milrila, unii-ju9yax04c7
PZRHRDRVRGEVNW-UHFFFAOYSA-N
6-methyl-2-oxo-5-pyridin-4-yl-1H-pyridine-3-carbonitrile
4197
211.22
Milrinone
C12H9N3O
50mg
14,6197
Soluble in DMSO; Insoluble in water.
CC1=C(C=C(C(=O)N1)C#N)C2=CC=NC=C2
211.224
CHEBI:50693
≥98%
Product Suggestions

Product Suggestions

Videos
SDS Documents

Documents

Certificates
Promotions

Promotions

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.