accessibility menu, dialog, popup

UserName

New Feature

You can now browse the site in English or Français.

H3COHNH2

CAS RN 78603-93-7

IUPAC Name: 2-amino-1,1-diphenylpropan-1-ol
Synonyms: 2-amino-1,1-diphenylpropan-1-ol
Molecular Weight (g/mol): 227.31
Molecular Formula: C15H17NO
InChi Key: FMBMNSFOFOAIMZ-UHFFFAOYNA-N
SMILES: CC(N)C(O)(C1=CC=CC=C1)C1=CC=CC=C1

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results