accessibility menu, dialog, popup

UserName

New Feature

You can now browse the site in English or Français.

H3COHO

CAS RN 79-09-4

IUPAC Name: propanoic acid
Synonyms: propanoic acid toxi-check caswell no. 707 propcorn propkorn fema number 2924 pseudoacetic acid C3 acid metacetonic acid kyselina propionova
Molecular Weight (g/mol): 74.08
Molecular Formula: C3H6O2
InChi Key: XBDQKXXYIPTUBI-UHFFFAOYSA-N
SMILES: CCC(O)=O
Molecular Weight (g/mol) 
  • (13)
  • (9)
Quantity 
  • (3)
  • (1)
  • (1)
  • (2)
  • (2)
  • (1)
  • (3)
  • (1)
  • (1)
  • (1)
  • (4)
Boiling Point 
  • (1)
  • (3)
  • (3)
  • (2)
  • (6)
Percent Purity 
  • (2)
  • (2)
  • (1)
  • (2)
  • (5)
  • (2)
Physical Form 
  • (12)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (1)
  • (3)
  • (2)
  • (1)
  • (1)
  • (1)
  • (1)
  • (7)
  • (3)
  • (2)
  • (13)
  • (9)
  • (3)
  • (1)
  • (1)
  • (2)
  • (2)
  • (1)
  • (3)
  • (1)
  • (1)
  • (1)
  • (4)
  • (1)
  • (3)
  • (3)
  • (2)
  • (6)
  • (2)
  • (2)
  • (1)
  • (2)
  • (5)
  • (2)
  • (12)
  • (1)
  • (2)
  • (2)
  • (4)
  • (1)
  • (1)
  • (1)