missing translation for 'onlineSavingsMsg'
Learn More

Propionic Acid, TRC

Product Code. 18083010
Change view
Click to view available options
Quantity:
10 g
100 g
Unit Size:
100g
10g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
18083010 100 g 100g
17909719 10 g 10g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 18083010 Supplier Toronto Research Chemicals Supplier No. TRCP786360100G

Please to purchase this item. Need a web account? Register with us today!

This item requires a price update prior to being ordered.

Request a Price for this item here.
, 02 950 59 478

This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 79-09-4
Molecular Formula C3 H6 O2
Molecular Weight (g/mol) 74.08
Synonym Propionic Acid, Propanoic acid, Propionic acid (6CI,8CI), Adofeed, Antischim B, Carboxyethane, Ethanecarboxylic acid, Ethylformic acid, Luprosil, Metacetonic acid, Methylacetic acid, MonoProp, Propcorn, Propkorn, Prozoin, Pseudoacetic acid, Toxi-Check
IUPAC Name propanoic acid
SMILES CCC(=O)O

Specifications

Quantity 100 g
InChI Formula InChI=1S/C3H6O2/c1-2-3(4)5/h2H2,1H3,(H,4,5)
Formula Weight 74.0368
Percent Purity >95%
Chemical Name or Material Propionic acid (Propanoic acid)
Grade GC
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.