accessibility menu, dialog, popup

UserName

New Feature

You can now browse the site in English or Français.

CAS RN 79-15-2

CH3ONHBr

CAS RN 79-15-2

IUPAC Name: N-bromoacetamide
Synonyms: acetamide, N-bromo-
Molecular Weight (g/mol): 137.96
Molecular Formula: C2H4BrNO
InChi Key: VBTQNRFWXBXZQR-UHFFFAOYSA-N
SMILES: CC(=O)NBr
Molecular Weight (g/mol) 
  • (3)
  • (3)
Quantity 
  • (1)
  • (1)
  • (1)
  • (2)
  • (1)
Percent Purity 
  • (1)
  • (2)
Physical Form 
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (1)
  • (2)
  • (3)
  • (3)
  • (3)
  • (1)
  • (1)
  • (1)
  • (2)
  • (1)
  • (1)
  • (2)
  • (1)