missing translation for 'onlineSavingsMsg'
Learn More

N-bromoacetamide, TRC

Product Code. 30336799
Change view
Click to view available options
Quantity:
10 g
Unit Size:
10g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
30336799 10 g 10g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 30336799 Supplier Toronto Research Chemicals Supplier No. TRCB68013410G
 Restricted Item
Estimated Shipment: 13-11-2026
Add to basket
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Specifikationer

Chemical Name or Material N-bromoacetamide
CAS 79-15-2
Molecular Formula C2H4BrNO
Quantity 10 g
Synonym Acetobromamide, NBA
InChI Formula InChI=1S/C2H4BrNO/c1-2(5)4-3/h1H3,(H,4,5)
SMILES CC(=O)NBr
IUPAC Name N-bromoacetamide
Molecular Weight (g/mol) 137.96
Formula Weight 136.9476
Produkttitel
Välj ett problem

Genom att klicka på Skicka bekräftar du att du kan bli kontaktad av Fisher Scientific angående feedbacken du har lämnat i detta formulär. Vi kommer inte att dela din information för andra ändamål. All kontaktinformation som tillhandahålls ska också underhållas i enlighet med vår Sekretesspolicy.