accessibility menu, dialog, popup

UserName

New Feature

You can now browse the site in English or Français.

CAS RN 79-43-6

IUPAC Name: 2,2-dichloroacetic acid
Synonyms: dichloroacetic acid dichloressigsaeure urner liquid DCA urner's liquid bichloracetic acid kyselina dichloroctova dca (acid) C2H2cl2O2
Molecular Weight (g/mol): 128.94
Molecular Formula: C2H2Cl2O2
InChi Key: JXTHNDFMNIQAHM-UHFFFAOYSA-N
SMILES: OC(=O)C(Cl)Cl
Molecular Weight (g/mol) 
  • (5)
  • (9)
Quantity 
  • (2)
  • (1)
  • (1)
  • (1)
  • (2)
  • (1)
  • (1)
  • (1)
  • (1)
  • (1)
  • (1)
Percent Purity 
  • (2)
  • (3)
  • (2)
  • (3)
Physical Form 
  • (3)
  • (7)
Boiling Point 
  • (2)
  • (4)
  • (3)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (2)
  • (1)
  • (1)
  • (3)
  • (4)
  • (3)
  • (5)
  • (9)
  • (2)
  • (1)
  • (1)
  • (1)
  • (2)
  • (1)
  • (1)
  • (1)
  • (1)
  • (1)
  • (1)
  • (2)
  • (3)
  • (2)
  • (3)
  • (3)
  • (7)
  • (2)
  • (4)
  • (3)