Learn More
Dichloroacetic Acid, Extra Pure, SLR, Fisher Chemical™
Product Code. 10283230 Shop All Fisher Chemical Products
Change view
Click to view available options
Quantity:
100 mL
1 L
Unit Size:
1 L
100mL
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
10283230 100 mL 100mL
10274342 1 L 1 L
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Product Code. 10283230 Supplier Fisher Chemical Supplier No. D/1550/PB05
This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products
missing translation for 'validMainframeMsg'
Chemical Identifiers
CAS 79-43-6
Molecular Formula C2H2Cl2O2
Molecular Weight (g/mol) 128.94
MDL Number 4223
InChI Key JXTHNDFMNIQAHM-UHFFFAOYSA-N
IUPAC Name 2,2-dichloroacetic acid
SMILES OC(=O)C(Cl)Cl
Specifications
Melting Point 9°C
Color Yellow
pH 1.2
Boiling Point 194°C
Packaging Plastic Coated Glass bottle
Quantity 100 mL
Vapor Density 4.45
Formula Weight 128.94g/mol
Vapor Pressure 1.3mbar 44
Physical Form Liquid
compliance-icons compliance-icons
Product Identifier
  • Dichloroacetic acid
Signal Word
  • Danger
Hazard Category
  • ACUTE AQUATIC Acute 1
  • Serious eye damage/eye irritation Category 1
  • Skin corrosion/irritation Category 1 A
Hazard Statement
  • H318-Causes serious eye damage.
  • H400-Very toxic to aquatic life.
  • H314-Causes severe skin burns and eye damage.
Precautionary Statement
  • P273-Avoid release to the environment.
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P301+P330+P331-IF SWALLOWED: rinse mouth. Do NOT induce vomiting.
  • P302+P352-IF ON SKIN: Wash with plenty of water/.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
  • P312-Call a POISON CENTER/doctor/if you feel unwell.