CAS RN 796738-71-1
IUPAC Name:
—
Synonyms:
—
Molecular Weight (g/mol):
—
Molecular Formula:
—
InChi Key:
—
SMILES:
—
1
–
3
of
3
results
1
N-(2-Methyl-4-nitrophenyl)-4-(3-pyridinyl)-2-pyrimidinamine, TRC
CAS: 796738-71-1 Molecular Formula: C16 H13 N5 O2 Molecular Weight (g/mol): 307.31 Synonym: 2-Pyrimidinamine, N-(2-methyl-4-nitrophenyl)-4-(3-pyridinyl)-,N-(2-Methyl-4-nitrophenyl)-4-(3-pyridinyl)-2-pyrimidinamine,N-(2-Methyl-4-nitrophenyl)-4-(3-pyridyl)-2-pyrimidineamine IUPAC Name: N-(2-methyl-4-nitrophenyl)-4-pyridin-3-ylpyrimidin-2-amine SMILES: Cc1cc(ccc1Nc2nccc(n2)c3cccnc3)[N+](=O)[O-]
3
N-(2-Methyl-4-nitrophenyl)-4-(3-pyridinyl)-2-pyrimidinamine, Mikromol™
Discover Mikromol - your trusted source for high-quality pharmaceutical reference standards. Supporting accurate, compliant analysis with ISO 17034-accredited materials designed for confidence in every result.