missing translation for 'onlineSavingsMsg'
Learn More

N-(2-Methyl-4-nitrophenyl)-4-(3-pyridinyl)-2-pyrimidinamine, TRC

Product Code. 30240629
Change view
Click to view available options
Quantity:
100 mg
500 mg
Unit Size:
100mg
500mg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
30240629 500 mg 500mg
30240621 100 mg 100mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 30240629 Supplier Toronto Research Chemicals Supplier No. TRCM325590500MG
 Restricted Item
Estimated Shipment: 10-09-2026
Add to basket
Add to basket
This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 796738-71-1
Molecular Formula C16 H13 N5 O2
Molecular Weight (g/mol) 307.31
Synonym 2-Pyrimidinamine, N-(2-methyl-4-nitrophenyl)-4-(3-pyridinyl)-, N-(2-Methyl-4-nitrophenyl)-4-(3-pyridinyl)-2-pyrimidinamine, N-(2-Methyl-4-nitrophenyl)-4-(3-pyridyl)-2-pyrimidineamine
IUPAC Name N-(2-methyl-4-nitrophenyl)-4-pyridin-3-ylpyrimidin-2-amine
SMILES Cc1cc(ccc1Nc2nccc(n2)c3cccnc3)[N+](=O)[O-]

Specifications

Quantity 500 mg
InChI Formula InChI=1S/C16H13N5O2/c1-11-9-13(21(22)23)4-5-14(11)19-16-18-8-6-15(20-16)12-3-2-7-17-10-12/h2-10H,1H3,(H,18,19,20)
Formula Weight 307.1069
Chemical Name or Material N-(2-Methyl-4-nitrophenyl)-4-(3-pyridinyl)-2-pyrimidinamine
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.