accessibility menu, dialog, popup

UserName

New Feature

You can now browse the site in English or Français.

OBrO

CAS RN 79839-49-9

IUPAC Name: 2-bromobenzene-1,3-dicarbaldehyde
Synonyms: 2-bromobenzene-1,3-dicarbaldehyde
Molecular Weight (g/mol): 213.03
Molecular Formula: C8H5BrO2
InChi Key: RZUSSKMZLHKMHU-UHFFFAOYSA-N
SMILES: BrC1=C(C=O)C=CC=C1C=O
Quantity 
  • (2)
  • (1)
  • (1)
  • (2)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results