accessibility menu, dialog, popup

UserName

New Feature

You can now browse the site in English or Français.

NNH3CClCH3

CAS RN 79917-90-1

IUPAC Name: 1-butyl-3-methyl-1H-imidazol-3-ium
Synonyms: 1-butyl-3-methylimidazolium N-butyl-N'-methylimidazolium
Molecular Weight (g/mol): 139.22
Molecular Formula: C8H15N2
InChi Key: IQQRAVYLUAZUGX-UHFFFAOYSA-N
SMILES: CCCCN1C=C[N+](C)=C1
Quantity 
  • (1)
  • (1)
  • (1)
  • (1)
  • (1)
  • (2)
  • (1)
  • (1)
Percent Purity 
  • (2)
  • (4)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (3)
  • (1)
  • (1)
  • (1)
  • (1)
  • (1)
  • (2)
  • (1)
  • (1)
  • (2)
  • (4)