missing translation for 'onlineSavingsMsg'
Learn More

1-Butyl-3-methylimidazolium Chloride, TRC

Product Code. 30012615
Change view
Click to view available options
Quantity:
1 g
2.5 g
5 g
Unit Size:
1g
2.5g
5g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
30012615 5 g 5g
30010817 2.5 g 2.5g
30010657 1 g 1g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 30012615 Supplier Toronto Research Chemicals Supplier No. TRCB8108555G
 Restricted Item
Estimated Shipment: 13-11-2026
Add to basket
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 79917-90-1
Molecular Formula C8H15ClN2
Molecular Weight (g/mol) 174.67
Synonym 1H-Imidazolium, 3-butyl-1-methyl-, chloride (1:1) (ACI), 1H-Imidazolium, 1-butyl-3-methyl-, chloride (9CI), 1-Butyl-3-methyiimidazoiium chloride, 1-Butyl-3-methyl-1H-imidazolium chloride, 1-Butyl-3-methylimidazolium chloride, 1-Butyl-3-methylimidizolium chloride, 1-Methyl-3-butylimidazolium chloride, 1-n-Butyl-3-methylimidazolium chloride, 3-Butyl-1-methylimidazolium chloride, [BMIM]Cl, [C4Mim+][Cl-], [C4Mim]Cl, Basionics ST 70, LM 1039, N-Butyl-N'-methylimidazolium chloride
IUPAC Name 1-butyl-3-methylimidazol-3-ium;chloride
SMILES [Cl-].CCCC[n+]1ccn(C)c1

Specifications

Quantity 5 g
InChI Formula InChI=1S/C8H15N2.ClH/c1-3-4-5-10-7-6-9(2)8-10;/h6-8H,3-5H2,1-2H3;1H/q+1;/p-1
Formula Weight 174.0924
Chemical Name or Material 1-Butyl-3-methylimidazolium Chloride
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.