accessibility menu, dialog, popup

UserName

New Feature

You can now browse the site in English or Français.

CAS RN 79944-56-2

HClNNHOO

CAS RN 79944-56-2

IUPAC Name: hydrogen 2-(2,3-dihydro-1,4-benzodioxin-2-yl)-4,5-dihydro-1H-imidazole chloride
Synonyms: hydrogen idazoxan chloride
Molecular Weight (g/mol): 240.69
Molecular Formula: C11H13ClN2O2
InChi Key: MYUBYOVCLMEAOH-UHFFFAOYNA-N
SMILES: [H+].[Cl-].C1CN=C(N1)C1COC2=CC=CC=C2O1
Quantity 
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results