missing translation for 'onlineSavingsMsg'
Learn More

Idazoxan Hydrochloride, TRC

Product Code. 30399711
Change view
Click to view available options
Quantity:
100 mg
1 g
Unit Size:
100mg
1g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
30399711 100 mg 100mg
30393104 1 g 1g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 30399711 Supplier Toronto Research Chemicals Supplier No. TRCI506800100MG
 Restricted Item
Estimated Shipment: 13-11-2026
Add to basket
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 79944-56-2
Molecular Formula C11H13ClN2O2
Molecular Weight (g/mol) 240.69
Synonym 2-(2,3-Dihydro-1,4-benzodioxin-2-yl)-4,5-dihydro-1H-imidazole Hydrochloride, (+/-)-Idazoxan Monohydrochloride, RX 781094, RX 781094A
IUPAC Name 2-(2,3-dihydro-1,4-benzodioxin-3-yl)-4,5-dihydro-1H-imidazole;hydrochloride
SMILES Cl.C1CN=C(N1)C2COc3ccccc3O2

Specifications

Quantity 100 mg
InChI Formula InChI=1S/C11H12N2O2.ClH/c1-2-4-9-8(3-1)14-7-10(15-9)11-12-5-6-13-11;/h1-4,10H,5-7H2,(H,12,13);1H
Formula Weight 240.0666
Chemical Name or Material Idazoxan Hydrochloride
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.