accessibility menu, dialog, popup

UserName

New Feature

You can now browse the site in English or Français.

CAS RN 80-05-7

IUPAC Name: 4-[2-(4-hydroxyphenyl)propan-2-yl]phenol
Synonyms: bisphenol-A DIAN rikabanol 4,4'-dihydroxydiphenylpropane p,p'-isopropylidenebisphenol 4,4'-isopropylidenediphenol ucar bisphenol A p,p'-bisphenol A ipognox 88 bisphenol a, sodium salt
Molecular Weight (g/mol): 228.29
Molecular Formula: C15H16O2
InChi Key: IISBACLAFKSPIT-UHFFFAOYSA-N
SMILES: CC(C)(C1=CC=C(O)C=C1)C1=CC=C(O)C=C1
Percent Purity 
  • (3)
  • (3)
  • (4)
Quantity 
  • (1)
  • (1)
  • (1)
  • (1)
  • (1)
  • (1)
  • (1)
  • (1)
  • (1)
  • (1)
Boiling Point 
  • (3)
  • (4)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (1)
  • (3)
  • (1)
  • (3)
  • (3)
  • (4)
  • (1)
  • (1)
  • (1)
  • (1)
  • (1)
  • (1)
  • (1)
  • (1)
  • (1)
  • (1)
  • (3)
  • (4)
  • (4)
  • (1)
  • (1)
  • (1)