accessibility menu, dialog, popup

UserName

New Feature

You can now browse the site in English or Français.

CH3NOCH3SOOCl

CAS RN 80466-79-1

IUPAC Name: dimethyl-1,2-oxazole-4-sulfonyl chloride
Synonyms: dimethyl-1,2-oxazole-4-sulfonyl chloride
Molecular Weight (g/mol): 195.62
Molecular Formula: C5H6ClNO3S
InChi Key: GZLPFEYTAAXJCP-UHFFFAOYSA-N
SMILES: CC1=C(C(C)=NO1)S(Cl)(=O)=O
Quantity 
  • (1)
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results