missing translation for 'onlineSavingsMsg'
Learn More

3,5-Dimethylisoxazole-4-sulfonyl chloride, 98%, Thermo Scientific Chemicals

Product Code. 11424347
Change view
Click to view available options
Quantity:
1 g
5 g
25 g
Unit Size:
1g
25g
5g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
11424347 25 g 25g
11404347 1 g 1g
11414347 5 g 5g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 11424347 Supplier Thermo Scientific Alfa Aesar Supplier No. B20411.14

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products

This item is not returnable. View return policy

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 80466-79-1
Molecular Formula C5H6ClNO3S
Molecular Weight (g/mol) 195.62
MDL Number MFCD00052554
InChI Key GZLPFEYTAAXJCP-UHFFFAOYSA-N
Synonym 3,5-dimethylisoxazole-4-sulfonyl chloride, 3,5-dimethylisoxazole-4-sulfonylchloride, 3,5-dimethylisoxazole-4-sulphonyl chloride, dimethyl-1,2-oxazole-4-sulfonyl chloride, 3,5-dimethyl-isoxazole-4-sulfonyl chloride, 3,5-dimethyl-4-isoxazolesulfonyl chloride, 4-isoxazolesulfonyl chloride, 3,5-dimethyl, 3,5-dimethylisoxazol-4-yl chlorosulfone, 4-chlorosulfonyl, dmisc
PubChem CID 2736319
SMILES CC1=C(C(C)=NO1)S(Cl)(=O)=O

Specifications

Melting Point 37°C to 40°C
Quantity 25 g
UN Number UN3261
Sensitivity Moisture sensitive
IUPAC Name 3,5-dimethyl-1,2-oxazole-4-sulfonyl chloride
Formula Weight 195.62
Percent Purity 98%
Chemical Name or Material 3,5-Dimethylisoxazole-4-sulfonyl chloride

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.