CAS RN 8065-71-2
IUPAC Name:
—
Synonyms:
—
Molecular Weight (g/mol):
—
Molecular Formula:
—
InChi Key:
—
SMILES:
—
1
–
1
of
1
results
1
Gliftor, TRC
CAS: 8065-71-2 Molecular Formula: C6H12ClF3O2 Molecular Weight (g/mol): 208.61 Synonym: 1-Chloro-3-fluoro-2-propanol mixt. with 1,3-difluoro-2-propanol,1,3-Difluoro-2-propanol mixt. contg. IUPAC Name: 1,3-difluoropropan-2-ol--1-chloro-3-fluoropropan-2-ol (1/1) SMILES: FCC(O)CF.FCC(O)CCl