missing translation for 'onlineSavingsMsg'
Learn More

Gliftor, TRC

Product Code. 30134114
Change view
Click to view available options
Quantity:
250 mg
2.5 g
Unit Size:
2.5g
250mg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
30134114 2.5 g 2.5g
30005104 250 mg 250mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 30134114 Supplier Toronto Research Chemicals Supplier No. TRCG4100002.5G

Please to purchase this item. Need a web account? Register with us today!

This item requires a price update prior to being ordered.

Request a Price for this item here.

This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 8065-71-2
Molecular Formula C6H12ClF3O2
Molecular Weight (g/mol) 208.61
Synonym 1-Chloro-3-fluoro-2-propanol mixt. with 1,3-difluoro-2-propanol, 1,3-Difluoro-2-propanol mixt. contg.
IUPAC Name 1,3-difluoropropan-2-ol--1-chloro-3-fluoropropan-2-ol (1/1)
SMILES FCC(O)CF.FCC(O)CCl

Specifications

Quantity 2.5 g
InChI Formula InChI=1S/C3H6ClFO.C3H6F2O/c2*4-1-3(6)2-5/h2*3,6H,1-2H2
Formula Weight 208.05
Chemical Name or Material Gliftor
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.