accessibility menu, dialog, popup

UserName

New Feature

You can now browse the site in English or Français.

OHOHOO

CAS RN 81-64-1

IUPAC Name: 1,4-dihydroxy-9,10-dihydroanthracene-9,10-dione
Synonyms: quinizarin quinizarine solvent orange 86 smoke orange R chinizarin macrolex orange GG
Molecular Weight (g/mol): 240.21
Molecular Formula: C14H8O4
InChi Key: GUEIZVNYDFNHJU-UHFFFAOYSA-N
SMILES: OC1=CC=C(O)C2=C1C(=O)C1=CC=CC=C1C2=O
Molecular Weight (g/mol) 
  • (6)
  • (3)
Quantity 
  • (1)
  • (1)
  • (1)
  • (1)
  • (1)
  • (2)
  • (1)
  • (1)
Boiling Point 
  • (3)
  • (3)
Percent Purity 
  • (3)
  • (3)
Physical Form 
  • (3)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (3)
  • (3)
  • (3)
  • (6)
  • (3)
  • (1)
  • (1)
  • (1)
  • (1)
  • (1)
  • (2)
  • (1)
  • (1)
  • (3)
  • (3)
  • (3)
  • (3)
  • (3)