accessibility menu, dialog, popup

UserName

New Feature

You can now browse the site in English or Français.

CAS RN 81228-09-3

IUPAC Name: 2-(2,4-difluorophenyl)acetate
Synonyms: 2-(2,4-difluorophenyl)acetate
Molecular Weight (g/mol): 171.12
Molecular Formula: C8H5F2O2
InChi Key: QPKZIGHNRLZBCL-UHFFFAOYSA-M
SMILES: [O-]C(=O)CC1=CC=C(F)C=C1F
Molecular Weight (g/mol) 
  • (4)
  • (3)
Quantity 
  • (2)
  • (2)
  • (3)
Percent Purity 
  • (1)
  • (1)
  • (2)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (1)
  • (3)
  • (4)
  • (3)
  • (2)
  • (2)
  • (3)
  • (1)
  • (1)
  • (2)
  • (2)