Learn More
2,4-Difluorobenzeneacetic Acid, TRC
Product Code. 30383849
Change view
Click to view available options
Quantity:
1 g
5 g
25 g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity
30383849 1 g
30374313 25 g
30382994 5 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Product Code. 30383849 Supplier Toronto Research Chemicals Supplier No. TRCD4454051G
 Restricted Item
Estimated Shipment: 10-12-2026
Add to basket
Add to basket
missing translation for 'validMainframeMsg'
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
TRUSTED_SUSTAINABILITY
Chemical Identifiers
CAS 81228-09-3
Molecular Formula C8H6F2O2
Molecular Weight (g/mol) 172.13
Synonym 2,4-Difluorophenylacetic Acid, 2-(2,4-Difluorophenyl)acetic Acid
IUPAC Name 2-(2,4-difluorophenyl)acetic acid
SMILES OC(=O)Cc1ccc(F)cc1F
Specifications
Quantity 1 g
InChI Formula InChI=1S/C8H6F2O2/c9-6-2-1-5(3-8(11)12)7(10)4-6/h1-2,4H,3H2,(H,11,12)
Formula Weight 172.0336
Chemical Name or Material 2,4-Difluorobenzeneacetic Acid