accessibility menu, dialog, popup

UserName

New Feature

You can now browse the site in English or Français.

CAS RN 814-89-1

CoOOOO

CAS RN 814-89-1

IUPAC Name: λ²-cobalt(2+) oxalate
Synonyms: cobalt(II) oxalate
Molecular Weight (g/mol): 146.95
Molecular Formula: C2CoO4
InChi Key: MULYSYXKGICWJF-UHFFFAOYSA-L
SMILES: [Co++].[O-]C(=O)C([O-])=O

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results