accessibility menu, dialog, popup

UserName

New Feature

You can now browse the site in English or Français.

CAS RN 81452-54-2

CH3SOOCH3

CAS RN 81452-54-2

IUPAC Name: methyl 3-methylthiophene-2-carboxylate
Synonyms: methyl 3-methylthiophene-2-carboxylate
Molecular Weight (g/mol): 156.2
Molecular Formula: C7H8O2S
InChi Key: BRWROFVPMUPMJQ-UHFFFAOYSA-N
SMILES: COC(=O)C1=C(C)C=CS1
Boiling Point 
  • (2)
Quantity 
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results