missing translation for 'onlineSavingsMsg'
Learn More

Methyl 3-methylthiophene-2-carboxylate, 99%

Product Code. 11334915
Change view
Click to view available options
Quantity:
1 g
5 g
Unit Size:
1g
5g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
11334915 5 g 5g
11324915 1 g 1g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 11334915 Supplier Thermo Scientific Alfa Aesar Supplier No. L18093.06
 Restricted Item
Estimated Shipment: 30-07-2026
Add to basket
This item is not returnable. View return policy

CAS: 81452-54-2 | C7H8O2S | 156.20 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 81452-54-2
Molecular Formula C7H8O2S
Molecular Weight (g/mol) 156.20
MDL Number MFCD00234332
InChI Key BRWROFVPMUPMJQ-UHFFFAOYSA-N
Synonym methyl 3-methyl-2-thiophenecarboxylate, 3-methylthiophene-2-carboxylic acid methyl ester, 3-methyl-thiophene-2-carboxylic acid methyl ester, 2-thiophenecarboxylic acid, 3-methyl-, methyl ester, methyl 3-methyl-thiophene-2-carboxylate, d methyl ester, 3-methyl-thiophene-2-carboxylic acid, pubchem10098, pubchem22915, ksc495k9b
PubChem CID 580757
IUPAC Name methyl 3-methylthiophene-2-carboxylate
SMILES COC(=O)C1=C(C)C=CS1

Specifications

Boiling Point 216°C to 217°C
Refractive Index 1.5168
Quantity 5 g
Beilstein 116836
Formula Weight 156.2
Percent Purity 99%
Chemical Name or Material Methyl 3-methylthiophene-2-carboxylate

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.