accessibility menu, dialog, popup

UserName

New Feature

You can now browse the site in English or Français.

H3CH3CCH3OOHO

CAS RN 815-17-8

IUPAC Name: 3,3-dimethyl-2-oxobutanoic acid
Synonyms: 3,3-dimethyl-2-oxobutanoic acid
Molecular Weight (g/mol): 130.14
Molecular Formula: C6H10O3
InChi Key: IAWVHZJZHDSEOC-UHFFFAOYSA-N
SMILES: CC(C)(C)C(=O)C(O)=O
Quantity 
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results