accessibility menu, dialog, popup

UserName

New Feature

You can now browse the site in English or Français.

ONOClClClClCl

CAS RN 82-68-8

IUPAC Name: 1,2,3,4,5-pentachloro-6-nitrobenzene
Synonyms: pentachloronitrobenzene PCNB quintozene nitropentachlorobenzene PKhNC
Molecular Weight (g/mol): 295.32
Molecular Formula: C6Cl5NO2
InChi Key: LKPLKUMXSAEKID-UHFFFAOYSA-N
SMILES: [O-][N+](=O)C1=C(Cl)C(Cl)=C(Cl)C(Cl)=C1Cl
Molecular Weight (g/mol) 
  • (11)
  • (2)
Solvent or Matrix 
  • (1)
  • (2)
  • (2)
  • (1)
  • (1)
  • (3)
  • (1)
Concentration 
  • (2)
  • (4)
  • (3)
Quantity 
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (1)
  • (8)
  • (1)
  • (2)
  • (1)
  • (1)
  • (11)
  • (2)
  • (1)
  • (2)
  • (2)
  • (1)
  • (1)
  • (3)
  • (1)
  • (2)
  • (4)
  • (3)
  • (1)
  • (1)