Learn More
Pentachloronitrobenzene 1000 μg/mL in Acetone, Dr. Ehrenstorfer
Product Code. 30310525
Change view
Click to view available options
Quantity:
1 mL
Unit Size:
1mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
30310525 1 mL 1mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Product Code. 30310525 Supplier Dr. Ehrenstorfer Supplier No. DREGA09011009AC
 Restricted Item
Estimated Shipment: 07-01-2027
Add to basket
Add to basket
missing translation for 'validMainframeMsg'
Discover Dr. Ehrenstorfer’s certified reference materials: available in multiple formats, including multi-component regulatory mixtures, to power your food and environmental analysis with traceable, ISO-accredited quality
TRUSTED_SUSTAINABILITY
Specifications
Chemical Name or Material Quintozene
CAS 82-68-8
Analyte or Component Names Quintozene
Shipping Condition Room Temperature
Molecular Formula C6 Cl5 N O2
Concentration 1000 μg/mL
Certifications/Compliance ISO 17034
Synonym Quintozene, Pentachloronitrobenzene, Benzene, 1,2,3,4,5-pentachloro-6-nitro-, Benzene, pentachloronitro- (8CI,9CI), 1,2,3,4,5-Pentachloro-6-nitrobenzene, 1,2,3,4,5-Pentachloronitrobenzene, 2,3,4,5,6-Pentachloronitrobenzene, Avicol (pesticide), Batrilex, Botrilex, Brassicol, Brassicol 75, Brassicol Super, Chinozan, Fartox, Folosan, Fomac 2, GC 3944-3-4, Kobutol, Korsikol, Marisan forte, NSC 58427, Nitropentachlorobenzene, PCNB, Pentagen, Phomasan, Plantacol, Quinosan, Quintozen, RTU 1010, RTU PCNB, Terrachlor, Terraclor, Terraclor 30G, Terrafun, Terrazan, Terrazan F, Tilcarex, Tritisan
InChI Formula InChI=1S/C6Cl5NO2/c7-1-2(8)4(10)6(12(13)14)5(11)3(1)9
SMILES [O-][N+](=O)c1c(Cl)c(Cl)c(Cl)c(Cl)c1Cl
IUPAC Name 1,2,3,4,5-pentachloro-6-nitrobenzene
Solvent or Matrix Acetone
Molecular Weight (g/mol) 295.33
Quantity 1 mL
Formula Weight 292.8372 g/mol