accessibility menu, dialog, popup

UserName

New Feature

You can now browse the site in English or Français.

CAS RN 821-09-0

HOCH2

CAS RN 821-09-0

IUPAC Name: pent-4-en-1-ol
Synonyms: 4-penten-1-ol
Molecular Weight (g/mol): 86.13
Molecular Formula: C5H10O
InChi Key: LQAVWYMTUMSFBE-UHFFFAOYSA-N
SMILES: OCCCC=C
Boiling Point 
  • (5)
Molecular Weight (g/mol) 
  • (2)
  • (3)
Percent Purity 
  • (3)
  • (2)
Quantity 
  • (1)
  • (1)
  • (1)
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (2)
  • (3)
  • (5)
  • (2)
  • (3)
  • (3)
  • (2)
  • (5)
  • (1)
  • (1)
  • (1)
  • (1)
  • (1)