accessibility menu, dialog, popup

UserName

New Feature

You can now browse the site in English or Français.

CAS RN 82257-09-8

IUPAC Name: 3-bromo-4-methoxypyridine
Synonyms: 3-bromo-4-methoxypyridine
Molecular Weight (g/mol): 188.02
Molecular Formula: C6H6BrNO
InChi Key: DSFJKFHIMBVWAV-UHFFFAOYSA-N
SMILES: COC1=C(Br)C=NC=C1
Quantity 
  • (1)
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results