CAS RN 82938-05-4
IUPAC Name:
—
Synonyms:
—
Molecular Weight (g/mol):
—
Molecular Formula:
—
InChi Key:
—
SMILES:
—
1
–
1
of
1
results
1
17Beta-Estradiol-13C2, TRC
CAS: 82938-05-4 Molecular Formula: 13C2 C16 H24 O2 Molecular Weight (g/mol): 274.37 Synonym: (17β)-Estra-1,3,5(10)-triene-3,17-diol-3,4-13C2,17β-Estradiol-3,4-13C2 IUPAC Name: (8R,9S,13S,14S,17S)-13-methyl-6,7,8,9,11,12,14,15,16,17-decahydrocyclopenta[a]phenanthrene-3,17-diol SMILES: C[C@]12CC[C@H]3[C@@H](CCc4[13cH][13c](O)ccc34)[C@@H]1CC[C@@H]2O