accessibility menu, dialog, popup

UserName

New Feature

You can now browse the site in English or Français.

H3COOCH3CH3OOCH3

CAS RN 83-27-2

IUPAC Name: 1,3-diethyl 2-(butan-2-yl)propanedioate
Synonyms: 1,3-diethyl 2-(sec-butyl)propanedioate
Molecular Weight (g/mol): 216.28
Molecular Formula: C11H20O4
InChi Key: MIIZSUOEOUHAIZ-UHFFFAOYNA-N
SMILES: CCOC(=O)C(C(C)CC)C(=O)OCC
Molecular Weight (g/mol) 
  • (3)
  • (2)
Quantity 
  • (1)
  • (1)
  • (1)
  • (1)
  • (1)
Boiling Point 
  • (2)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results