accessibility menu, dialog, popup

UserName

New Feature

You can now browse the site in English or Français.

CH3H3CNOO

CAS RN 83-41-0

IUPAC Name: 1,2-dimethyl-3-nitrobenzene
Synonyms: 1,2-dimethyl-3-nitrobenzene
Molecular Weight (g/mol): 151.17
Molecular Formula: C8H9NO2
InChi Key: FVHAWXWFPBPFOS-UHFFFAOYSA-N
SMILES: CC1=CC=CC(=C1C)[N+]([O-])=O

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results