missing translation for 'onlineSavingsMsg'
Learn More

3-Nitro-o-xylene, 99%

Product Code. 11441407
Change view
Click to view available options
Quantity:
100 g
500 g
Unit Size:
100g
500g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
11441407 500 g 500g
11431407 100 g 100g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 11441407 Supplier Thermo Scientific Alfa Aesar Supplier No. B24917.36
 Restricted Item
In Stock
Add to basket
This item is not returnable. View return policy

CAS: 83-41-0 | C8H9NO2 | 151.17 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 83-41-0
Molecular Formula C8H9NO2
Molecular Weight (g/mol) 151.17
MDL Number MFCD00007162
InChI Key FVHAWXWFPBPFOS-UHFFFAOYSA-N
Synonym 3-nitro-o-xylene, 2,3-dimethylnitrobenzene, nitroxylene, o-xylene, 3-nitro, benzene, 1,2-dimethyl-3-nitro, xylene, ar-nitro, benzene, dimethylnitro, unii-49gnx4cy5w, ccris 3117, 49gnx4cy5w
PubChem CID 6739
IUPAC Name 1,2-dimethyl-3-nitrobenzene
SMILES CC1=CC=CC(=C1C)[N+]([O-])=O

Specifications

Melting Point 13°C to 15°C
Density 1.129
Quantity 500 g
Refractive Index 1.544
UN Number UN1665
Beilstein 2045105
Boiling Point 244°C to 246°C
Formula Weight 151.17
Percent Purity 99%
Chemical Name or Material 3-Nitro-o-xylene

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.