accessibility menu, dialog, popup

UserName

New Feature

You can now browse the site in English or Français.

OHH2NFFFFFFH2NOH

CAS RN 83558-87-6

IUPAC Name: 2-amino-4-[2-(3-amino-4-hydroxyphenyl)-1,1,1,3,3,3-hexafluoropropan-2-yl]phenol
Synonyms: 2-amino-4-[2-(3-amino-4-hydroxyphenyl)-1,1,1,3,3,3-hexafluoropropan-2-yl]phenol
Molecular Weight (g/mol): 366.26
Molecular Formula: C15H12F6N2O2
InChi Key: MSTZGVRUOMBULC-UHFFFAOYSA-N
SMILES: NC1=C(O)C=CC(=C1)C(C1=CC(N)=C(O)C=C1)(C(F)(F)F)C(F)(F)F
Molecular Weight (g/mol) 
  • (3)
  • (2)
Quantity 
  • (2)
  • (1)
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results