missing translation for 'onlineSavingsMsg'
Learn More

2,2-Bis(3-amino-4-hydroxyphenyl)hexafluoropropane, 97%, Thermo Scientific™

Product Code. 15595916 Shop All Thermo Scientific Chemicals Products
Change view
Click to view available options
Quantity:
1 g
5 g
Unit Size:
1g
5g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
15595916 1 g 1g
15505926 5 g 5g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 15595916 Supplier Thermo Scientific Acros Supplier No. 456400010

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products

This item is not returnable. View return policy

Chemical Identifiers

CAS 83558-87-6
Molecular Formula C15H12F6N2O2
Molecular Weight (g/mol) 366.27
InChI Key MSTZGVRUOMBULC-UHFFFAOYSA-N
Synonym 2,2-bis 3-amino-4-hydroxyphenyl hexafluoropropane, 4,4'-perfluoropropane-2,2-diyl bis 2-aminophenol, 2,2-bis 3-amino-4-hydroxyphenyl-hexafluoropropane, 2-amino-4-2-3-amino-4-hydroxyphenyl-1,1,1,3,3,3-hexafluoropropan-2-yl phenol, 4,4'-hexafluoroisopropylidene bis 2-aminophenol, phenol, 4,4'-2,2,2-trifluoro-1-trifluoromethyl ethylidene bis 2-amino, 4,4'-1,1,1,3,3,3-hexafluoropropane-2,2-diyl bis 2-aminophenol, 2,2-bis 3-amino-4-hydrophenyl hexafluoropropane, 1,1-bis 3-amino-4-hydroxyphenyl-2,2,2-trifluoro-1-trifluoromethyl ethane
PubChem CID 632603
IUPAC Name 2-amino-4-[2-(3-amino-4-hydroxyphenyl)-1,1,1,3,3,3-hexafluoropropan-2-yl]phenol
SMILES C1=CC(=C(C=C1C(C2=CC(=C(C=C2)O)N)(C(F)(F)F)C(F)(F)F)N)O

Specifications

Melting Point 238.0°C to 240.0°C
Infrared Spectrum Authentic
Assay Percent Range 96% min. (HPLC)
Packaging Glass Bottle
Quantity 1 g
Formula Weight 366.27
Percent Purity 97%
Chemical Name or Material 2,2-Bis(3-amino-4-hydroxyphenyl)hexafluoropropane
compliance-icons
Product Identifier
  • 2,2-Bis(3-amino-4-hydroxyphenyl) hexafluoropropane, 97%
Signal Word
  • Warning
Hazard Category
  • Serious eye damage/eye irritation Category 2
  • Skin corrosion/irritation Category 2
  • Specific target organ toxicity Category 3
Hazard Statement
  • H319-Causes serious eye irritation.
  • H335-May cause respiratory irritation.
  • H315-Causes skin irritation.
Precautionary Statement
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P302+P352-IF ON SKIN: Wash with plenty of water/.
  • P304+P340-IF INHALED: Remove person to fresh air and keep comfortable for breathing.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
  • P312-Call a POISON CENTER/doctor/if you feel unwell.

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.