accessibility menu, dialog, popup

UserName

New Feature

You can now browse the site in English or Français.

CAS RN 83643-88-3

CH3(S)H2NOHOOHNO

CAS RN 83643-88-3

IUPAC Name: (2S)-2-amino-3-(5-methyl-3-oxo-2,3-dihydro-1,2-oxazol-4-yl)propanoic acid
Synonyms: (2S)-2-amino-3-(5-methyl-3-oxo-2H-1,2-oxazol-4-yl)propanoic acid
Molecular Weight (g/mol): 186.17
Molecular Formula: C7H10N2O4
InChi Key: UUDAMDVQRQNNHZ-YFKPBYRVSA-N
SMILES: CC1=C(C[C@H](N)C(O)=O)C(=O)NO1

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results