missing translation for 'onlineSavingsMsg'
Learn More

(S)-alpha-Amino-3-hydroxy-5-methyl-4-isoxazolepropionic acid, Thermo Scientific™

Product Code. 15400487
Click to view available options
Quantity:
5 mg
Unit Size:
5mg
This item is not returnable. View return policy

Product Code. 15400487

Brand: Thermo Scientific Alfa Aesar J65847.#0

Request Bulk or Custom Format

Please to purchase this item. Need a web account? Register with us today!

This item has been discontinued and is no longer available. View the product for possible alternatives or contact our Technical Support team on 056 260 260 for assistance.
View alternative products

This item is not returnable. View return policy

Active enantiomer of AMPA that induces apoptosis in cortical neurons

This Thermo Scientific brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.

TRUSTED_SUSTAINABILITY

Specifications

CAS 83643-88-3
Color White
Molecular Formula C7H10N2O4
MDL Number MFCD00672630
Quantity 5 mg
Sensitivity Hygroscopic
Synonym s-ampa, s-alpha-amino-3-hydroxy-5-methyl-4-isoxazolepropionic acid, ampa, l, unii-9280sc28gd, s-2-amino-3-3-hydroxy-5-methyl-isoxazol-4-yl-propionic acid, amq, glutamate-ampa, 1ftm, tocris-0169, tocris-0254
InChI Key UUDAMDVQRQNNHZ-YFKPBYRVSA-N
SMILES CC1=C(C(=O)NO1)CC(C(=O)O)N
IUPAC Name (2S)-2-amino-3-(5-methyl-3-oxo-1,2-oxazol-4-yl)propanoic acid
Molecular Weight (g/mol) 186.167
PubChem CID 158397
Formula Weight 186.16
Physical Form Powder
Chemical Name or Material (S)-alpha-Amino-3-hydroxy-5-methyl-4-isoxazolepropionic acid
Show More Show Less
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.