accessibility menu, dialog, popup

UserName

New Feature

You can now browse the site in English or Français.

H3COOONOO

CAS RN 838-57-3

IUPAC Name: ethyl 3-(4-nitrophenyl)-3-oxopropanoate
Synonyms: ethyl 3-(4-nitrophenyl)-3-oxopropanoate
Molecular Weight (g/mol): 237.21
Molecular Formula: C11H11NO5
InChi Key: NGRXSVFCLHVGKU-UHFFFAOYSA-N
SMILES: CCOC(=O)CC(=O)C1=CC=C(C=C1)[N+]([O-])=O
Quantity 
  • (1)
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results