accessibility menu, dialog, popup

UserName

New Feature

You can now browse the site in English or Français.

CAS RN 84-54-8

H3COO

CAS RN 84-54-8

IUPAC Name: 2-methyl-9,10-dihydroanthracene-9,10-dione
Synonyms: 2-methylanthraquinone tectochinon 2-methylanthra-9,10-quinone tectoquinone techtoquinone
Molecular Weight (g/mol): 222.24
Molecular Formula: C15H10O2
InChi Key: NJWGQARXZDRHCD-UHFFFAOYSA-N
SMILES: CC1=CC=C2C(=O)C3=CC=CC=C3C(=O)C2=C1
Percent Purity 
  • (1)
  • (3)
Quantity 
  • (1)
  • (1)
  • (1)
  • (1)
Physical Form 
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (1)
  • (3)
  • (1)
  • (1)
  • (3)
  • (1)
  • (1)
  • (1)
  • (1)
  • (1)