accessibility menu, dialog, popup

UserName

New Feature

You can now browse the site in English or Français.

CAS RN 84-65-1

IUPAC Name: 9,10-dihydroanthracene-9,10-dione
Synonyms: anthraquinone anthradione morkit corbit ccris 649 caswell no. 052A anthrapel epa pesticide chemical code 122701 bis-alkylamino anthraquinone 9,10-quinone
Molecular Weight (g/mol): 208.22
Molecular Formula: C14H8O2
InChi Key: RZVHIXYEVGDQDX-UHFFFAOYSA-N
SMILES: O=C1C2=CC=CC=C2C(=O)C2=CC=CC=C12
Molecular Weight (g/mol) 
  • (4)
  • (8)
Quantity 
  • (1)
  • (1)
  • (1)
  • (3)
  • (1)
  • (1)
Boiling Point 
  • (2)
  • (3)
Percent Purity 
  • (2)
  • (3)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (1)
  • (1)
  • (3)
  • (4)
  • (8)
  • (1)
  • (1)
  • (1)
  • (3)
  • (1)
  • (1)
  • (2)
  • (3)
  • (2)
  • (3)
  • (3)
  • (2)
  • (1)
  • (1)
  • (1)