accessibility menu, dialog, popup

UserName

New Feature

You can now browse the site in English or Français.

H3CCH3OOOOCH3CH3

CAS RN 84-69-5

IUPAC Name: 1,2-bis(2-methylpropyl) benzene-1,2-dicarboxylate
Synonyms: diisobutyl phthalate DIBP hatcol dibp kodaflex dibp di(i-butyl)phthalate hexaplas M/1B di-isobutylphthalate diisobutylester kyseliny ftalove palatinol IC
Molecular Weight (g/mol): 278.35
Molecular Formula: C16H22O4
InChi Key: MGWAVDBGNNKXQV-UHFFFAOYSA-N
SMILES: CC(C)COC(=O)C1=CC=CC=C1C(=O)OCC(C)C
Quantity 
  • (1)
  • (1)
  • (1)
  • (1)
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results