accessibility menu, dialog, popup

UserName

New Feature

You can now browse the site in English or Français.

H3COOOOCH3

CAS RN 84-76-4

IUPAC Name: 1,2-dinonyl benzene-1,2-dicarboxylate
Synonyms: dinonyl phthalate
Molecular Weight (g/mol): 418.62
Molecular Formula: C26H42O4
InChi Key: DROMNWUQASBTFM-UHFFFAOYSA-N
SMILES: CCCCCCCCCOC(=O)C1=CC=CC=C1C(=O)OCCCCCCCCC
Quantity 
  • (1)
  • (2)
  • (1)
  • (1)
  • (1)
Percent Purity 
  • (2)
  • (3)
Boiling Point 
  • (2)
Physical Form 
  • (2)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (1)
  • (1)
  • (2)
  • (3)
  • (1)
  • (2)
  • (1)
  • (1)
  • (1)
  • (2)
  • (3)
  • (2)
  • (2)