missing translation for 'onlineSavingsMsg'
Learn More

Dinonyl Phthalate, MP Biomedicals

Product Code. 11408354 Shop All MP Biomedicals Products
Change view
Click to view available options
Quantity:
100 g
Unit Size:
25g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
11408354 100 g 25g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 11408354 Supplier MP Biomedicals Supplier No. 0521073325
 Restricted Item
Estimated Shipment: 19-08-2026
Add to basket
Add to basket
This item is not returnable. View return policy

Specifications

CAS 84-76-4
Molecular Formula C26H42O4
MDL Number MFCD00036237
Quantity 100 g
Synonym dinonyl phthalate, dinonylphthalate, nonyl phthalate, di-n-nonyl phthalate, bisoflex dnp, unimoll dn, bisoflex 91, phthalic acid, dinonyl ester, 1,2-benzenedicarboxylic acid, dinonyl ester, bisolflex 91
InChI Key DROMNWUQASBTFM-UHFFFAOYSA-N
SMILES CCCCCCCCCOC(=O)C1=CC=CC=C1C(=O)OCCCCCCCCC
IUPAC Name 1,2-dinonyl benzene-1,2-dicarboxylate
Molecular Weight (g/mol) 418.62
PubChem CID 6787
Formula Weight 418.62
Chemical Name or Material Dinonyl Phthalate
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.