CAS RN 84225-95-6
IUPAC Name:
3,5-dichloro-N-{[(2R)-1-ethylpyrrolidin-2-yl]methyl}-2-hydroxy-6-methoxybenzamide
Synonyms:
raclopride
Molecular Weight (g/mol):
347.24
Molecular Formula:
C15H20Cl2N2O3
InChi Key:
WAOQONBSWFLFPE-SECBINFHSA-N
SMILES:
CCN1CCC[C@@H]1CNC(=O)C1=C(O)C(Cl)=CC(Cl)=C1OC
1
–
3
of
3
results
1
Raclopride, TRC
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
2
Tocris Bioscience™ Raclopride
CAS: 84225-95-6 Molecular Formula: C15H20Cl2N2O3
3
Raclopride, TRC
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.