CAS RN 84746-24-7
IUPAC Name:
—
Synonyms:
—
Molecular Weight (g/mol):
—
Molecular Formula:
—
InChi Key:
—
SMILES:
—
1
–
2
of
2
results
1
2,2'-(Piperazine-1,4-diyl)dipyrimidine, Mikromol™
Discover Mikromol - your trusted source for high-quality pharmaceutical reference standards. Supporting accurate, compliant analysis with ISO 17034-accredited materials designed for confidence in every result.
2
2,2’-(1,4-Piperazinediyl)bis-pyrimidine, TRC
CAS: 84746-24-7 Molecular Formula: C12 H14 N6 Molecular Weight (g/mol): 242.28 Synonym: 2,2'-(Piperazine-1,4-diyl)dipyrimidine,Buspirone Hydrochloride Imp. G (EP) IUPAC Name: 2-(4-pyrimidin-2-ylpiperazin-1-yl)pyrimidine SMILES: C1CN(CCN1c2ncccn2)c3ncccn3