missing translation for 'onlineSavingsMsg'
Learn More

2,2’-(1,4-Piperazinediyl)bis-pyrimidine, TRC

Product Code. 30133006
Change view
Click to view available options
Quantity:
500 mg
5 g
Unit Size:
500mg
5g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
30133006 5 g 5g
30010101 500 mg 500mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 30133006 Supplier Toronto Research Chemicals Supplier No. TRCP9917705G
 Restricted Item
Estimated Shipment: 13-11-2026
Add to basket
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 84746-24-7
Molecular Formula C12 H14 N6
Molecular Weight (g/mol) 242.28
Synonym 2,2'-(Piperazine-1,4-diyl)dipyrimidine, Buspirone Hydrochloride Imp. G (EP)
IUPAC Name 2-(4-pyrimidin-2-ylpiperazin-1-yl)pyrimidine
SMILES C1CN(CCN1c2ncccn2)c3ncccn3

Specifications

Quantity 5 g
InChI Formula InChI=1S/C12H14N6/c1-3-13-11(14-4-1)17-7-9-18(10-8-17)12-15-5-2-6-16-12/h1-6H,7-10H2
Formula Weight 242.128
Percent Purity >95%
Chemical Name or Material 2,2'-(Piperazine-1,4-diyl)dipyrimidine
Grade HPLC
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.